Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 5 | 524 | ||
XM_005271642.2 | XP_005271699.1 | 4 | 413 | |||
XM_005271643.2 | XP_005271700.1 | 2 | 230 | |||
XM_006718894.2 | XP_006718957.1 | 5 | 472 | |||
XM_011542944.2 | XP_011541246.1 | 5 | 609 | |||
XM_011542945.2 | XP_011541247.1 | 6 | 569 | |||
XM_011542946.1 | XP_011541248.1 | 5 | 492 | |||
XM_011542947.2 | XP_011541249.1 | 4 | 350 | |||
XM_017018110.1 | XP_016873599.1 | 5 | 3359 | |||
XM_017018111.1 | XP_016873600.1 | 4 | 391 | |||
XM_017018112.1 | XP_016873601.1 | 2 | 230 | |||
GenBank mRNA | AB065001.1 | - | 2 | 202 | ||
AJ417081.1 | - | 2 | 174 | |||
AK023136.1 | - | 2 | 202 | |||
AK295857.1 | - | 4 | 391 | |||
AK316001.1 | - | 4 | 324 | |||
AK316268.1 | - | 3 | 312 | |||
BC045626.1 | - | 5 | 481 | 5261106 | ||
BX648615.1 | - | 3 | 301 | |||
DA789677.1 | - | 3 | 346 |