Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 15 | 2917 | ||
XM_005257567.1 | XP_005257624.1 | 15 | 2904 | |||
XM_017024934.1 | XP_016880423.1 | 15 | 2829 | |||
XM_017024935.1 | XP_016880424.1 | 15 | 2826 | |||
GenBank mRNA | AB051521.1 | - | 15 | 2917 | ||
AK022940.1 | - | 2 | 415 | |||
AK025999.1 | - | 7 | 1396 | |||
AK225412.1 | - | 7 | 1396 | |||
AL832432.1 | - | 8 | 1842 | |||
BC022237.1 | - | 2 | 435 | |||
BC025977.1 | - | 2 | 316 | |||
BC036820.1 | - | 10 | 2053 | |||
BC051868.2 | - | 7 | 1260 | 6162673 |