Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022068.3 | NP_071351.2 | 44 | 7094 | ||
XM_011525723.2 | XP_011524025.1 | 47 | 8024 | |||
XM_011525724.2 | XP_011524026.1 | 46 | 7967 | |||
XM_011525725.2 | XP_011524027.1 | 45 | 7934 | |||
XM_011525726.2 | XP_011524028.1 | 46 | 7841 | |||
XM_017025918.1 | XP_016881407.1 | 46 | 7985 | |||
GenBank mRNA | AK027056.1 | - | 4 | 501 | ||
AK302764.1 | - | 6 | 931 | |||
JN790819.1 | - | 44 | 6928 |