Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303472.1 | NP_001290401.1 | 2 | 484 | ||
NM_022096.5 | NP_071379.3 | 3 | 548 | |||
NM_198798.2 | NP_942093.1 | 2 | 484 | |||
GenBank mRNA | AK097689.1 | - | 2 | 417 | ||
BC022878.2 | - | 3 | 458 | 4821619 | ||
BI826033.1 | - | 2 | 435 | |||
BP279314.1 | - | 2 | 484 |