Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040283.2 | NP_001035373.1 | 3, 4 | 460 | ||
XM_011527214.1 | XP_011525516.1 | 3, 4 | 432 | |||
XM_011527215.1 | XP_011525517.1 | 3, 4 | 424 | |||
XM_011527216.2 | XP_011525518.1 | 3, 4 | 479 | |||
XM_017027123.1 | XP_016882612.1 | 3, 4 | 477 | |||
XM_017027124.1 | XP_016882613.1 | 3, 4 | 479 | |||
XM_017027125.1 | XP_016882614.1 | 3, 4 | 482 | |||
XM_017027126.1 | XP_016882615.1 | 3, 4 | 572 | |||
XM_017027127.1 | XP_016882616.1 | 3, 4 | 437 | |||
XM_017027128.1 | XP_016882617.1 | 3, 4 | 642 | |||
XM_017027129.1 | XP_016882618.1 | 3, 4 | 473 | |||
GenBank mRNA | AJ291669.1 | - | 2, 3 | 276 | ||
AK057404.1 | - | 3, 4 | 424 | |||
AK301822.1 | - | 3, 4 | 463 | |||
BC029202.1 | - | 3, 4 | 426 | 5170924 | ||
BC039266.1 | - | 3, 4 | 428 | 5298983 | ||
DB026313.1 | - | 3, 4 | 463 |