Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022728.3 | NP_073565.2 | 2 | 1020 | ||
GenBank mRNA | AF063609.1 | - | 2 | 823 | ||
AF136169.1 | - | 2 | 788 | |||
AF248954.1 | - | 2 | 1020 | |||
AK055224.1 | - | 1 | 2117 | |||
AK055238.1 | - | 2 | 990 | |||
AK289619.1 | - | 2 | 821 | |||
BC035048.2 | - | 2 | 822 | 5261681 |