Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137605.2 | NP_001131077.1 | 9, 10 | 1011 | ||
NM_022141.6 | NP_071424.1 | 9, 10 | 1071 | |||
XM_005261702.3 | XP_005261759.1 | 9, 10 | 798 | |||
XM_011530302.2 | XP_011528604.1 | 8, 9 | 3563 | |||
XM_017028907.1 | XP_016884396.1 | 9, 10 | 2009 | |||
XM_017028908.1 | XP_016884397.1 | 9, 10 | 2029 | |||
GenBank mRNA | AF237772.1 | - | 9, 10 | 865 | ||
AK225929.1 | - | 10, 11 | 961 | |||
AL590887.1 | - | 7, 8 | 587 | |||
BC034406.1 | - | 9, 10 | 992 | 5178529 | ||
BX648942.1 | - | 4, 5 | 3928 | |||
CA440742.1 | - | 5, 6 | 578 | |||
CR456480.2 | - | 8, 9 | 642 |