Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPFFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022146.4 | NP_071429.1 | 3 | 356 | ||
GenBank mRNA | AB040104.1 | - | 2 | 338 | ||
AF268898.1 | - | 2 | 338 | |||
AF330055.1 | - | 2 | 338 | |||
BC131580.1 | - | 2 | 338 | 40112416 | ||
EU432128.1 | - | 2 | 338 |
Set Membership: |
G-protein coupled receptor |