Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022167.3 | NP_071450.2 | 5 | 1130 | ||
NR_110010.1 | 5 | 1130 | ||||
XM_005257572.4 | XP_005257629.1 | 5 | 1444 | |||
GenBank mRNA | AJ277442.1 | - | 5 | 1130 | ||
AK299999.1 | - | 5 | 1027 | |||
AY358090.1 | - | 5 | 1026 | |||
BC052262.1 | - | 5 | 1050 | 6380013 |