Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2BPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024496.3 | NP_078772.1 | 1 | 942 | ||
GenBank mRNA | AB058768.1 | - | 1 | 522 | ||
AI821684.1 | - | 1 | 237 | |||
BC108292.1 | - | 1 | 942 | |||
BC152431.1 | - | 1 | 522 | |||
BM744266.1 | - | 1 | 110 |