Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022361.4 | NP_071756.2 | 2 | 543 | ||
XM_011536067.2 | XP_011534369.1 | 2 | 703 | |||
XM_017011194.1 | XP_016866683.1 | 2 | 1986 | |||
XM_017011195.1 | XP_016866684.1 | 2 | 1446 | |||
GenBank mRNA | AF204171.1 | - | 1 | 288 | ||
AK314101.1 | - | 2 | 543 | |||
BC022323.2 | - | 1 | 393 | 4293961 |