Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022370.3 | NP_071765.2 | 9, 10 | 1696 | ||
XM_017018122.1 | XP_016873611.1 | 8, 9 | 1461 | |||
GenBank mRNA | AK291045.1 | - | 9, 10 | 1630 | ||
AY509035.1 | - | 9, 10 | 1719 |