Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022370.3 | NP_071765.2 | 25 | 3928 | ||
XM_017018122.1 | XP_016873611.1 | 24 | 3693 | |||
GenBank mRNA | AK024697.1 | - | 9 | 1165 | ||
AK225076.1 | - | 9 | 1165 | |||
AK291045.1 | - | 25 | 3862 | |||
AK298792.1 | - | 8 | 1028 | |||
AY509035.1 | - | 25 | 3951 | |||
BC008623.1 | - | 5 | 905 |