Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022453.2 | NP_071898.2 | 10 | 1085 | ||
XM_017004695.1 | XP_016860184.1 | 10 | 1085 | |||
GenBank mRNA | AK023968.1 | - | 10 | 1003 | ||
AK222525.1 | - | 10 | 1023 | |||
AK289501.1 | - | 10 | 1029 | |||
BC015612.2 | - | 10 | 1002 | 4811222 |