Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278712.1 | NP_001265641.1 | 1, 2 | 517 | ||
NM_022488.4 | NP_071933.2 | 1, 2 | 517 | |||
XM_011513074.1 | XP_011511376.1 | 1, 2 | 127 | |||
GenBank mRNA | AA448974.1 | - | 1, 2 | 478 | ||
AB079384.1 | - | 1, 2 | 82 | |||
AF202092.1 | - | 1, 2 | 97 | |||
AK307432.1 | - | 1, 2 | 67 | |||
AK308919.1 | - | 1, 2 | 478 | |||
AL137515.1 | - | 1, 2 | 217 | |||
AW408464.1 | - | 1, 2 | 424 | |||
BC002830.1 | - | 1, 2 | 475 | 3633903 | ||
BC024221.2 | - | 1, 2 | 192 | 3938447 | ||
DC311625.1 | - | 1, 2 | 517 |