Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZDHHC6 SIRNA Assays ›
Gene Symbol:
ZDHHC6
Gene Name:
zinc finger DHHC-type containing 6
Gene Aliases:
DHHC-6, ZNF376
Chromosome Location:
Chr.10: 112424411 - 112448335 on Build GRCh38
UniGene ID:
Hs.196990
Species:
Human
siRNA ID
HSS127620
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZDHHC6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303134.1
NP_001290063.1
9, 10
1545
NM_022494.2
NP_071939.1
9, 10
1557
XM_005270051.4
XP_005270108.1
9, 10
2492
XM_006717948.3
XP_006718011.1
9, 10
1663
XM_006717949.3
XP_006718012.1
8, 9
1392
XM_011540091.2
XP_011538393.1
9, 10
1545
XM_011540092.2
XP_011538394.1
9, 10
1462
XM_011540093.1
XP_011538395.1
9, 10
1571
XM_011540094.2
XP_011538396.1
9, 10
1430
XM_011540095.2
XP_011538397.1
8, 9
1280
XM_017016561.1
XP_016872050.1
9, 10
1483
XM_017016562.1
XP_016872051.1
9, 10
1651
XM_017016563.1
XP_016872052.1
9, 10
1464
XM_017016564.1
XP_016872053.1
9, 10
1663
XM_017016565.1
XP_016872054.1
9, 10
1729
XM_017016566.1
XP_016872055.1
9, 10
1542
XM_017016567.1
XP_016872056.1
8, 9
1380
XM_017016568.1
XP_016872057.1
9, 10
1368
XM_017016569.1
XP_016872058.1
5, 6
505
GenBank mRNA
AF267740.1
-
9, 10
1511
AK025605.1
-
9, 10
1512
AK026369.1
-
4, 5
417
AK223086.1
-
9, 10
1511
BC007213.1
-
9, 10
1487
2960984
BC017434.2
-
9, 10
1487
2960984
BX647270.1
-
7, 8
4001
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0