Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033225.5 | NP_150094.5 | 70 | 10981 | ||
XM_011534752.2 | XP_011533054.1 | 69 | 11165 | |||
XM_011534753.2 | XP_011533055.1 | 53 | 8088 | |||
XM_011534754.1 | XP_011533056.1 | 46 | 6824 | |||
GenBank mRNA | AB209502.1 | - | 56 | 8799 | ||
AF333704.2 | - | 71 | 10987 | |||
AK127129.1 | - | 20 | 2954 | |||
AY017307.1 | - | 68 | 10351 |