Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033225.5 | NP_150094.5 | 3 | 709 | ||
XM_011534752.2 | XP_011533054.1 | 3 | 938 | |||
XM_017013731.1 | XP_016869220.1 | 3 | 938 | |||
GenBank mRNA | AF333704.2 | - | 3 | 709 | ||
AY017307.1 | - | 3 | 604 |