Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 10 | 1206 | ||
GenBank mRNA | AK021562.1 | - | 3 | 245 | ||
AK304372.1 | - | 9 | 854 | |||
AY700779.1 | - | 2 | 134 | |||
BC069665.1 | - | 3 | 223 | |||
BC069726.1 | - | 3 | 226 | |||
BC069747.1 | - | 3 | 225 | |||
BC069768.1 | - | 3 | 226 | |||
BC074794.2 | - | 2 | 162 | |||
BC113478.1 | - | 2 | 190 | |||
BC113504.1 | - | 2 | 190 |