Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166426.1 | NP_001159898.1 | 5 | 1697 | ||
NM_017883.4 | NP_060353.2 | 6 | 1362 | |||
NR_029427.1 | 6 | 1328 | ||||
XM_006724539.1 | XP_006724602.1 | 7 | 1015 | |||
XM_006724541.3 | XP_006724604.1 | 7 | 981 | |||
XM_011543947.2 | XP_011542249.1 | 7 | 975 | |||
GenBank mRNA | AF158978.1 | - | 5 | 1724 | ||
AF329819.4 | - | 6 | 1362 | |||
AK314739.1 | - | 6 | 1325 | |||
BC002507.2 | - | 3 | 424 | |||
BC067094.1 | - | 6 | 1009 | |||
BC080579.1 | - | 6 | 1091 | 6644248 | ||
BX649101.1 | - | 4 | 307 | |||
DQ907252.1 | - | 7 | 1054 | |||
DQ907253.1 | - | 5 | 1369 | |||
DQ907254.1 | - | 5 | 1363 | |||
DQ907255.1 | - | 5 | 1312 |