Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166426.1 | NP_001159898.1 | 7 | 2063 | ||
NM_017883.4 | NP_060353.2 | 8 | 1728 | |||
NR_029427.1 | 8 | 1694 | ||||
XM_006724539.1 | XP_006724602.1 | 9 | 1381 | |||
XM_006724541.3 | XP_006724604.1 | 9 | 1347 | |||
XM_011543947.2 | XP_011542249.1 | 9 | 1341 | |||
GenBank mRNA | AF158978.1 | - | 7 | 2090 | ||
AF329819.4 | - | 8 | 1728 | |||
AK000570.1 | - | 3 | 374 | |||
AK225542.1 | - | 3 | 374 | |||
AK314739.1 | - | 8 | 1691 | |||
BC002507.2 | - | 5 | 790 | |||
BC067094.1 | - | 8 | 1375 | |||
BC080579.1 | - | 8 | 1457 | 6644248 | ||
BX649101.1 | - | 6 | 673 | |||
DQ907252.1 | - | 9 | 1420 | |||
DQ907253.1 | - | 7 | 1735 | |||
DQ907254.1 | - | 7 | 1729 | |||
DQ907255.1 | - | 7 | 1678 |