Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022746.3 | NP_073583.3 | 2 | 539 | ||
XM_011509900.2 | XP_011508202.1 | 2 | 539 | |||
XM_011509903.2 | XP_011508205.1 | 2 | 915 | |||
XM_011509904.2 | XP_011508206.1 | 2 | 192 | |||
XM_017002096.1 | XP_016857585.1 | 2 | 119 | |||
XM_017002097.1 | XP_016857586.1 | 2 | 172 | |||
GenBank mRNA | AK026043.1 | - | 2 | 336 | ||
AK225416.1 | - | 2 | 336 | |||
AK290812.1 | - | 2 | 311 | |||
BC010619.1 | - | 2 | 307 | 3872779 |