Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163315.2 | NP_001156787.2 | 3 | 1639 | ||
XM_005272048.4 | XP_005272105.1 | 3 | 1633 | |||
XM_005272050.4 | XP_005272107.1 | 3 | 1633 | |||
XM_011543574.2 | XP_011541876.1 | 3 | 1633 | |||
XM_011543575.2 | XP_011541877.1 | 3 | 1633 | |||
XM_011543576.2 | XP_011541878.1 | 3 | 1633 | |||
XM_011543577.2 | XP_011541879.1 | 3 | 1633 | |||
XM_011543578.2 | XP_011541880.1 | 3 | 1633 | |||
XM_011543579.2 | XP_011541881.1 | 3 | 1633 | |||
XM_011543580.2 | XP_011541882.1 | 3 | 1633 | |||
XM_017009729.1 | XP_016865218.1 | 3 | 1633 | |||
GenBank mRNA | AB533280.1 | - | 4 | 559 | ||
AK126722.1 | - | 4 | 827 | |||
AL133602.1 | - | 2 | 240 | |||
BC126144.1 | - | 3 | 365 | 8991860 | ||
BC126146.1 | - | 3 | 365 | 8991862 |