Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022828.3 | NP_073739.3 | 23 | 3429 | ||
XM_011543581.1 | XP_011541883.1 | 23 | 3440 | |||
XM_011543582.2 | XP_011541884.1 | 23 | 3414 | |||
XM_011543586.2 | XP_011541888.1 | 23 | 3414 | |||
XM_017009730.1 | XP_016865219.1 | 23 | 3440 | |||
XM_017009731.1 | XP_016865220.1 | 22 | 3215 | |||
XM_017009732.1 | XP_016865221.1 | 22 | 3215 | |||
XM_017009733.1 | XP_016865222.1 | 21 | 3043 | |||
XM_017009734.1 | XP_016865223.1 | 21 | 3043 | |||
XM_017009735.1 | XP_016865224.1 | 20 | 2862 | |||
GenBank mRNA | AK025593.1 | - | 7 | 1042 | ||
AK131450.1 | - | 22 | 3122 | |||
AK225316.1 | - | 7 | 1042 | |||
BC137285.1 | - | 23 | 3379 | 9021282 | ||
BE295472.1 | - | 3 | 443 |