Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022897.4 | NP_075048.1 | 22, 23 | 2576 | ||
XM_011534627.1 | XP_011532929.1 | 22, 23 | 2444 | |||
XM_011534631.2 | XP_011532933.1 | 21, 22 | 2445 | |||
XM_017009736.1 | XP_016865225.1 | 21, 22 | 2324 | |||
XM_017009737.1 | XP_016865226.1 | 21, 22 | 2457 | |||
XM_017009738.1 | XP_016865227.1 | 21, 22 | 2297 | |||
XM_017009739.1 | XP_016865228.1 | 21, 22 | 2285 | |||
XM_017009740.1 | XP_016865229.1 | 20, 21 | 2165 | |||
XM_017009741.1 | XP_016865230.1 | 22, 23 | 2444 | |||
XM_017009742.1 | XP_016865231.1 | 22, 23 | 2432 | |||
XM_017009743.1 | XP_016865232.1 | 22, 23 | 2618 | |||
XM_017009745.1 | XP_016865234.1 | 14, 15 | 1641 | |||
XM_017009747.1 | XP_016865236.1 | 10, 11 | 916 | |||
GenBank mRNA | AF222747.1 | - | 22, 23 | 2432 | ||
AJ271459.1 | - | 22, 23 | 2440 | |||
AJ288952.1 | - | 25, 26 | 2779 | |||
AJ288953.1 | - | 24, 25 | 2618 | |||
AJ288954.1 | - | 24, 25 | 2739 | |||
AJ288955.1 | - | 23, 24 | 2558 | |||
AK027880.1 | - | 20, 21 | 2260 | |||
AK057478.1 | - | 7, 8 | 771 | |||
AK315159.1 | - | 23, 24 | 2644 |