Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251969.1 | NP_001238898.1 | 3 | 1151 | ||
NM_022902.4 | NP_075053.2 | 4 | 605 | |||
NM_024055.4 | NP_076960.1 | 4 | 1274 | |||
XM_005248569.3 | XP_005248626.1 | 3 | 435 | |||
XM_006714672.3 | XP_006714735.1 | 4 | 599 | |||
XM_017009749.1 | XP_016865238.1 | 3 | 435 | |||
GenBank mRNA | AF461760.1 | - | 4 | 499 | ||
AK022818.1 | - | 2 | 88 | |||
AK292925.1 | - | 4 | 469 | |||
AY089991.1 | - | 4 | 298 | |||
AY358505.1 | - | 4 | 516 | |||
BC000808.1 | - | 4 | 1119 | |||
BC017441.1 | - | 4 | 1232 | |||
BX537394.1 | - | 4 | 298 |