Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134231.1 | NP_001127703.1 | 11, 12 | 1237 | ||
NM_022908.2 | NP_075059.1 | 11, 12 | 1510 | |||
XM_005265406.2 | XP_005265463.1 | 11, 12 | 1317 | |||
XM_006713303.3 | XP_006713366.1 | 11, 12 | 1231 | |||
XM_006713305.2 | XP_006713368.1 | 11, 12 | 1060 | |||
GenBank mRNA | AF131781.1 | - | 8, 9 | 779 | ||
AK022504.1 | - | 11, 12 | 1040 | |||
AK023995.1 | - | 11, 12 | 1497 | |||
AK092469.1 | - | 1, 2 | 1274 | |||
BC014550.1 | - | 10, 11 | 919 | 4286007 |