Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203365.3 | NP_976241.1 | 12 | 1720 | ||
NM_213589.2 | NP_998754.1 | 10 | 1564 | |||
XM_005246777.3 | XP_005246834.1 | 12 | 1715 | |||
XM_005246779.4 | XP_005246836.1 | 12 | 2614 | |||
XM_006712695.3 | XP_006712758.1 | 13 | 2695 | |||
XM_011511646.2 | XP_011509948.1 | 13 | 1787 | |||
XM_011511647.1 | XP_011509949.1 | 10 | 1420 | |||
XM_011511648.1 | XP_011509950.1 | 10 | 1414 | |||
XM_011511649.2 | XP_011509951.1 | 12 | 1715 | |||
GenBank mRNA | AB051468.1 | - | 9 | 1297 | ||
AB053311.1 | - | 12 | 1714 | |||
AJ584699.1 | - | 10 | 1558 | |||
AY494951.1 | - | 10 | 1560 | |||
AY523977.1 | - | 12 | 1716 | |||
AY523978.1 | - | 12 | 1716 |