Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171948.1 | NP_001165419.1 | 4 | 509 | ||
NM_001171949.1 | NP_001165420.1 | 3 | 413 | |||
NM_024069.3 | NP_076974.2 | 3 | 378 | |||
XM_005260073.4 | XP_005260130.1 | 3 | 352 | |||
XM_006722883.2 | XP_006722946.1 | 3 | 378 | |||
XM_011528272.2 | XP_011526574.1 | 3 | 1084 | |||
XM_011528273.2 | XP_011526575.1 | 3 | 583 | |||
XM_017027281.1 | XP_016882770.1 | 3 | 676 | |||
XM_017027282.1 | XP_016882771.1 | 3 | 676 | |||
GenBank mRNA | AF173381.1 | - | 3 | 261 | ||
AK091145.1 | - | 3 | 1668 | |||
AK098346.1 | - | 3 | 352 | |||
BC001080.2 | - | 3 | 346 | 2823374 | ||
BC006479.1 | - | 3 | 346 | 2823374 | ||
BM781878.1 | - | 3 | 296 | |||
BX648739.1 | - | 3 | 268 | |||
DA542130.1 | - | 4 | 424 | |||
DA616317.1 | - | 3 | 378 |