Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024078.2 | NP_076983.1 | 3 | 351 | ||
XM_017019960.1 | XP_016875449.1 | 3 | 356 | |||
GenBank mRNA | BC001191.1 | - | 3 | 330 | 3354945 | |
BC007893.2 | - | 3 | 223 | |||
BC071958.1 | - | 3 | 294 | 6597539 | ||
HY051629.1 | - | 3 | 352 |