Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270891.1 | NP_001257820.1 | 3 | 224 | ||
NM_001270892.1 | NP_001257821.1 | 2 | 198 | |||
NM_001270893.1 | NP_001257822.1 | 2 | 156 | |||
NM_024108.2 | NP_077013.1 | 3 | 266 | |||
XM_017027295.1 | XP_016882784.1 | 3 | 325 | |||
GenBank mRNA | AF161407.1 | - | 3 | 218 | ||
BC001907.1 | - | 3 | 250 | 3534111 | ||
BC004450.1 | - | 3 | 251 | 2820830 | ||
BI462720.1 | - | 2 | 155 | |||
CN256686.1 | - | 2 | 193 | |||
CN256687.1 | - | 3 | 236 | |||
FJ418644.1 | - | 3 | 206 |