Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003676.2 | NP_001003676.1 | 3, 4 | 404 | ||
NM_001003677.2 | NP_001003677.1 | 3, 4 | 404 | |||
NM_001003678.2 | NP_001003678.1 | 3, 4 | 404 | |||
NM_001278222.1 | NP_001265151.1 | 2, 3 | 301 | |||
NM_024113.4 | NP_077018.1 | 3, 4 | 404 | |||
XM_006718315.1 | XP_006718378.1 | 3, 4 | 395 | |||
XM_011520364.2 | XP_011518666.1 | 2, 3 | 466 | |||
XM_011520365.2 | XP_011518667.1 | 3, 4 | 395 | |||
XM_017018276.1 | XP_016873765.1 | 3, 4 | 395 | |||
GenBank mRNA | AK025863.1 | - | 3, 4 | 313 | ||
AK300813.1 | - | 2, 3 | 211 | |||
AK315639.1 | - | 3, 4 | 352 | |||
AL136575.1 | - | 3, 4 | 323 | |||
BC001860.2 | - | 3, 4 | 350 | 3534541 | ||
BC009455.1 | - | 3, 4 | 325 | 3535652 | ||
BC013988.2 | - | 3, 4 | 315 | 3349493 | ||
BC052291.1 | - | 3, 4 | 317 | 5936112 | ||
BQ428072.1 | - | 4, 5 | 584 | |||
CR457351.1 | - | 3, 4 | 287 | |||
DB027437.1 | - | 3, 4 | 395 |