Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024560.2 | NP_078836.1 | 13, 14 | 1806 | ||
XM_005269150.2 | XP_005269207.1 | 13, 14 | 1740 | |||
XM_011538740.2 | XP_011537042.1 | 13, 14 | 1455 | |||
GenBank mRNA | AK025616.1 | - | 13, 14 | 1757 | ||
AK074267.1 | - | 3, 4 | 230 | |||
AX764480.1 | - | 13, 14 | 1956 | |||
AX764482.1 | - | 13, 14 | 1712 | |||
BC009317.2 | - | 13, 14 | 1806 | 4120563 | ||
BC015769.1 | - | 13, 14 | 1806 | 4902942 |