Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142279.2 | NP_001135751.1 | 8 | 1044 | ||
NM_024570.3 | NP_078846.2 | 8 | 1044 | |||
XM_006719867.3 | XP_006719930.1 | 8 | 733 | |||
XM_011535229.1 | XP_011533531.1 | 8 | 966 | |||
XM_011535230.2 | XP_011533532.1 | 8 | 966 | |||
XM_011535231.2 | XP_011533533.1 | 8 | 966 | |||
XM_011535233.2 | XP_011533535.1 | 9 | 919 | |||
XM_017020747.1 | XP_016876236.1 | 8 | 966 | |||
GenBank mRNA | AK021774.1 | - | 8 | 931 | ||
AK124228.1 | - | 3 | 2710 | |||
AK223340.1 | - | 8 | 931 | |||
AY764036.1 | - | 8 | 952 | |||
BC001397.2 | - | 8 | 754 | |||
BC005088.1 | - | 8 | 754 | |||
BC007332.2 | - | 8 | 754 | |||
BC010174.2 | - | 8 | 754 | |||
BC036744.1 | - | 8 | 915 | 5195426 | ||
BX328027.1 | - | 5 | 371 | |||
BX330672.1 | - | 3 | 620 | |||
CA309004.1 | - | 3 | 638 |