Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253826.1 | NP_001240755.1 | 9, 10 | 1481 | ||
NM_001253827.1 | NP_001240756.1 | 10, 11 | 1246 | |||
NM_024572.3 | NP_078848.2 | 8, 9 | 1466 | |||
XM_011533105.1 | XP_011531407.1 | 10, 11 | 1000 | |||
XM_017004906.1 | XP_016860395.1 | 8, 9 | 931 | |||
XM_017004907.1 | XP_016860396.1 | 8, 9 | 942 | |||
XM_017004908.1 | XP_016860397.1 | 8, 9 | 1225 | |||
GenBank mRNA | AB078144.1 | - | 8, 9 | 826 | ||
AK022753.1 | - | 9, 10 | 902 | |||
AK074394.1 | - | 2, 3 | 126 | |||
AK091313.1 | - | 8, 9 | 836 | |||
AK122747.1 | - | 10, 11 | 1230 | |||
AK225808.1 | - | 8, 9 | 1189 | |||
AK298735.1 | - | 9, 10 | 894 | |||
AY358758.1 | - | 8, 9 | 1159 | |||
BC006269.2 | - | 1, 2 | 35 | |||
BC010659.1 | - | 8, 9 | 1466 | 3874588 | ||
Y09324.4 | - | 8, 9 | 826 |