Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040610.2 | NP_001035700.1 | 5, 6 | 750 | ||
NM_001322844.1 | NP_001309773.1 | 6, 7 | 818 | |||
NM_001322845.1 | NP_001309774.1 | 7, 8 | 1145 | |||
NM_024580.5 | NP_078856.4 | 7, 8 | 903 | |||
NR_136410.1 | 7, 8 | 903 | ||||
GenBank mRNA | AK023181.1 | - | 1, 2 | 55 | ||
AK056656.1 | - | 7, 8 | 833 | |||
AK300348.1 | - | 7, 8 | 813 | |||
BX538332.1 | - | 5, 6 | 681 |