Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024591.4 | NP_078867.2 | 3, 4 | 344 | ||
XM_005257668.1 | XP_005257725.1 | 3, 4 | 344 | |||
GenBank mRNA | AK021811.1 | - | 3, 4 | 282 | ||
AK223187.1 | - | 3, 4 | 306 | |||
AK292105.1 | - | 3, 4 | 310 | |||
AY329087.1 | - | 3, 4 | 266 | |||
BC010108.2 | - | 3, 4 | 290 | 3604039 | ||
BP196406.1 | - | 3, 4 | 344 | |||
CR457284.1 | - | 3, 4 | 266 |