Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167911.1 | NP_001161383.1 | 7 | 1210 | ||
NM_001167912.1 | NP_001161384.1 | 7 | 1210 | |||
NM_024621.2 | NP_078897.2 | 7 | 1401 | |||
XM_011513134.2 | XP_011511436.1 | 6 | 1144 | |||
XM_011513135.1 | XP_011511437.1 | 7 | 1040 | |||
GenBank mRNA | AB051479.1 | - | 7 | 1210 | ||
AK027625.1 | - | 7 | 1299 | |||
AL713656.1 | - | 6 | 998 | |||
BC101660.1 | - | 6 | 1037 | 8069166 | ||
BC111017.1 | - | 7 | 1202 | 5174889 | ||
BC113555.1 | - | 6 | 1037 | 8322607 |