Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253849.1 | NP_001240778.1 | 2 | 255 | ||
NM_001253850.1 | NP_001240779.1 | 2 | 173 | |||
NM_024626.3 | NP_078902.2 | 2 | 173 | |||
NR_045603.1 | 2 | 173 | ||||
NR_045604.1 | 2 | 173 | ||||
XM_011542143.2 | XP_011540445.2 | 2 | 457 | |||
XM_017002335.1 | XP_016857824.1 | 3 | 483 | |||
GenBank mRNA | AK026071.1 | - | 2 | 138 | ||
AK225413.1 | - | 2 | 140 | |||
AK303466.1 | - | 2 | 255 | |||
AK310812.1 | - | 2 | 140 | |||
AY280972.1 | - | 2 | 68 | |||
AY346100.1 | - | 2 | 128 | |||
AY358352.1 | - | 2 | 128 | |||
AY442303.1 | - | 2 | 141 | |||
BC065717.1 | - | 2 | 144 | 4295775 | ||
BC074729.2 | - | 2 | 99 | 30915182 | ||
BX648021.1 | - | 3 | 157 | |||
DB198367.1 | - | 2 | 140 | |||
DC347131.1 | - | 2 | 173 | |||
DQ103757.1 | - | 2 | 68 | |||
DQ836392.1 | - | 2 | 161 |