Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024701.3 | NP_078977.2 | 5 | 739 | ||
NR_024581.1 | 4 | 588 | ||||
NR_037164.1 | 6 | 820 | ||||
GenBank mRNA | AF403031.1 | - | 5 | 721 | ||
AK023196.1 | - | 5 | 722 | |||
AK091935.1 | - | 4 | 588 | |||
AK292071.1 | - | 5 | 725 | |||
BC012056.1 | - | 6 | 797 | 4552643 | ||
CR457302.1 | - | 5 | 695 |