Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324318.1 | NP_001311247.1 | 12 | 1557 | ||
NM_001324319.1 | NP_001311248.1 | 11 | 1437 | |||
NM_024745.4 | NP_079021.3 | 12 | 1668 | |||
NR_136738.1 | 13 | 1748 | ||||
GenBank mRNA | AK025662.1 | - | 8 | 974 | ||
AK055931.1 | - | 12 | 1639 | |||
AK225315.1 | - | 8 | 974 | |||
AK292164.1 | - | 12 | 1635 | |||
AK315706.1 | - | 12 | 1637 | |||
BC000960.1 | - | 7 | 752 | |||
BC030699.1 | - | 12 | 1643 | 4829528 |