Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136474.2 | NP_001129946.1 | 4 | 572 | ||
NM_001136475.2 | NP_001129947.1 | 2 | 288 | |||
NM_001301056.1 | NP_001287985.1 | 3 | 768 | |||
NM_024749.4 | NP_079025.2 | 3 | 768 | |||
XM_011509987.2 | XP_011508289.1 | 6 | 1052 | |||
XM_011509988.2 | XP_011508290.1 | 7 | 1126 | |||
XM_011509989.2 | XP_011508291.1 | 3 | 746 | |||
XM_011509991.2 | XP_011508293.2 | 2 | 465 | |||
XM_011509994.1 | XP_011508296.1 | 3 | 746 | |||
XM_017002351.1 | XP_016857840.1 | 4 | 886 | |||
XM_017002352.1 | XP_016857841.1 | 2 | 270 | |||
GenBank mRNA | AK022567.1 | - | 4 | 498 | ||
AK096605.1 | - | 4 | 436 | |||
AK290973.1 | - | 4 | 531 | |||
AK302675.1 | - | 2 | 141 | |||
AY834202.1 | - | 2 | 343 | |||
BC028194.1 | - | 2 | 167 | 5164251 | ||
BC051856.1 | - | 3 | 653 | 6067012 | ||
BC053836.1 | - | 3 | 669 | 5802700 |