Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284403.1 | NP_001271332.1 | 7 | 679 | ||
NM_001284404.1 | NP_001271333.1 | 8 | 809 | |||
NM_001284405.1 | NP_001271334.1 | 9 | 927 | |||
NM_024754.4 | NP_079030.3 | 10 | 1006 | |||
XM_005248601.2 | XP_005248658.1 | 9 | 886 | |||
XM_005248603.2 | XP_005248660.1 | 9 | 862 | |||
XM_017009866.1 | XP_016865355.1 | 9 | 867 | |||
XM_017009867.1 | XP_016865356.1 | 8 | 779 | |||
XM_017009868.1 | XP_016865357.1 | 8 | 748 | |||
GenBank mRNA | AK022660.1 | - | 8 | 768 | ||
AK056761.1 | - | 10 | 1670 | |||
AK298750.1 | - | 7 | 670 | |||
AK303639.1 | - | 8 | 778 | |||
BC018720.1 | - | 10 | 982 | 4552964 | ||
CR457290.1 | - | 6 | 534 |