Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145527.1 | NP_001138999.1 | 11 | 1702 | ||
NM_024757.4 | NP_079033.4 | 11 | 1702 | |||
XM_005266105.4 | XP_005266162.1 | 11 | 1745 | |||
XM_005266110.1 | XP_005266167.1 | 10 | 1617 | |||
XM_006717288.2 | XP_006717351.1 | 11 | 1663 | |||
XM_011519021.2 | XP_011517323.1 | 11 | 2700 | |||
XM_011519022.2 | XP_011517324.1 | 11 | 2155 | |||
XM_011519023.2 | XP_011517325.1 | 11 | 2679 | |||
XM_011519024.2 | XP_011517326.1 | 10 | 2623 | |||
XM_011519025.1 | XP_011517327.1 | 11 | 1865 | |||
XM_011519027.2 | XP_011517329.1 | 11 | 2701 | |||
XM_011519028.2 | XP_011517330.1 | 11 | 2701 | |||
XM_011519029.2 | XP_011517331.1 | 3 | 290 | |||
XM_017015134.1 | XP_016870623.1 | 11 | 2133 | |||
XM_017015135.1 | XP_016870624.1 | 11 | 1685 | |||
XM_017015136.1 | XP_016870625.1 | 10 | 1607 | |||
XM_017015137.1 | XP_016870626.1 | 11 | 1844 | |||
XM_017015138.1 | XP_016870627.1 | 10 | 1596 | |||
GenBank mRNA | AB028932.1 | - | 6 | 522 | ||
AB058779.2 | - | 6 | 522 | |||
AK022941.1 | - | 10 | 1617 | |||
AY083210.1 | - | 9 | 1572 |