Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK11 SIRNA Assays ›
Gene Symbol:
NEK11
Gene Name:
NIMA related kinase 11
Gene Aliases:
-
Chromosome Location:
Chr.3: 131026850 - 131350465 on Build GRCh38
UniGene ID:
Hs.657336
Species:
Human
siRNA ID
HSS129004
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146003.1
NP_001139475.1
13
1604
NM_001321220.1
NP_001308149.1
13
1604
NM_001321221.1
NP_001308150.1
15
1803
NM_001321222.1
NP_001308151.1
10
1289
NM_001321223.1
NP_001308152.1
13
1722
NM_001321224.1
NP_001308153.1
11
1330
NM_024800.4
NP_079076.3
14
1677
NM_145910.3
NP_665917.1
14
1677
XM_011513167.2
XP_011511469.1
14
1877
XM_011513173.1
XP_011511475.1
12
1353
XM_011513174.1
XP_011511476.1
14
1623
XM_011513175.1
XP_011511477.1
13
1465
XM_011513181.1
XP_011511483.1
10
1006
XM_017007205.1
XP_016862694.1
15
1980
XM_017007206.1
XP_016862695.1
15
1898
XM_017007207.1
XP_016862696.1
15
2114
XM_017007208.1
XP_016862697.1
14
1698
XM_017007209.1
XP_016862698.1
14
1907
XM_017007210.1
XP_016862699.1
14
1763
XM_017007211.1
XP_016862700.1
14
1698
XM_017007212.1
XP_016862701.1
13
1568
XM_017007213.1
XP_016862702.1
14
1676
XM_017007214.1
XP_016862703.1
13
1779
XM_017007215.1
XP_016862704.1
13
1568
XM_017007216.1
XP_016862705.1
12
1464
XM_017007217.1
XP_016862706.1
11
1361
XM_017007218.1
XP_016862707.1
16
1881
XM_017007219.1
XP_016862708.1
13
1375
XM_017007220.1
XP_016862709.1
14
1698
XM_017007221.1
XP_016862710.1
14
1698
XM_017007222.1
XP_016862711.1
14
1698
XM_017007223.1
XP_016862712.1
9
1149
XM_017007224.1
XP_016862713.1
15
1742
XM_017007225.1
XP_016862714.1
15
1755
XM_017007226.1
XP_016862715.1
15
2019
XM_017007227.1
XP_016862716.1
13
1499
XM_017007228.1
XP_016862717.1
11
1114
XM_017007229.1
XP_016862718.1
11
1361
XM_017007230.1
XP_016862719.1
11
1539
XM_017007231.1
XP_016862720.1
12
1369
XM_017007232.1
XP_016862721.1
14
1682
XM_017007233.1
XP_016862722.1
13
1432
GenBank mRNA
AB071996.1
-
14
1644
AB071997.1
-
14
1644
AK027148.1
-
9
870
AK293261.1
-
11
1245
AK297300.1
-
10
1289
AK316352.1
-
10
1204
AL833472.1
-
13
1575
BC028587.2
-
13
1721
4829802
BU674219.1
-
2
432
BX328072.2
-
7
672
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
histone phosphorylation
G2 DNA damage checkpoint
intra-S DNA damage checkpoint
intracellular signal transduction
mitotic cell cycle phase transition
regulation of mitotic cell cycle phase transition
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0