Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146254.1 | NP_001139726.1 | 8 | 1775 | ||
NM_001146255.1 | NP_001139727.1 | 6 | 1508 | |||
NM_024825.3 | NP_079101.3 | 8 | 1799 | |||
XM_017027313.1 | XP_016882802.1 | 7 | 1656 | |||
GenBank mRNA | AK027100.1 | - | 6 | 1499 | ||
AK290731.1 | - | 8 | 1732 | |||
AK296101.1 | - | 6 | 1495 | |||
AK298489.1 | - | 8 | 1775 | |||
BC057786.1 | - | 8 | 1799 | 30347370 |