Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP8B4 SIRNA Assays ›
Gene Symbol:
ATP8B4
Gene Name:
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases:
ATPIM
Chromosome Location:
Chr.15: 49858238 - 50182096 on Build GRCh38
UniGene ID:
Hs.511311
Species:
Human
siRNA ID
HSS129087
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP8B4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024837.3
NP_079113.2
12
997
NR_073596.1
13
1085
NR_073597.1
12
997
NR_073598.1
13
1104
XM_011522046.2
XP_011520348.1
14
1386
XM_011522047.2
XP_011520349.1
13
1911
XM_011522048.1
XP_011520350.1
13
1184
XM_011522049.2
XP_011520351.1
13
1294
XM_011522051.2
XP_011520353.1
14
1221
XM_011522052.2
XP_011520354.1
13
1279
XM_011522053.1
XP_011520355.1
13
1060
XM_011522056.2
XP_011520358.2
13
1446
XM_011522058.2
XP_011520360.1
9
1655
XM_011522059.1
XP_011520361.1
8
676
XM_011522060.1
XP_011520362.1
11
911
XM_011522061.1
XP_011520363.1
10
826
XM_011522062.1
XP_011520364.1
8
665
XM_011522063.1
XP_011520365.1
8
647
XM_011522064.1
XP_011520366.1
4
348
XM_011522069.2
XP_011520371.1
3
169
XM_017022587.1
XP_016878076.1
12
1362
XM_017022588.1
XP_016878077.1
12
1287
XM_017022589.1
XP_016878078.1
13
1446
XM_017022590.1
XP_016878079.1
14
1198
XM_017022591.1
XP_016878080.1
13
1145
XM_017022592.1
XP_016878081.1
12
1208
XM_017022593.1
XP_016878082.1
13
1446
XM_017022594.1
XP_016878083.1
9
706
XM_017022595.1
XP_016878084.1
9
2711
XM_017022596.1
XP_016878085.1
4
339
XM_017022597.1
XP_016878086.1
13
1446
XM_017022598.1
XP_016878087.1
13
1446
XM_017022599.1
XP_016878088.1
13
1446
GenBank mRNA
AB075819.1
-
13
1104
BC144297.1
-
12
993
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
phospholipid translocation
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0