Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271989.1 | NP_001258918.1 | 9 | 1455 | ||
NM_024899.3 | NP_079175.2 | 10 | 1680 | |||
NR_073537.1 | 10 | 1675 | ||||
XM_005258149.4 | XP_005258206.1 | 10 | 1659 | |||
XM_017025981.1 | XP_016881470.1 | 10 | 1659 | |||
XM_017025982.1 | XP_016881471.1 | 10 | 1740 | |||
XM_017025983.1 | XP_016881472.1 | 9 | 1530 | |||
XM_017025984.1 | XP_016881473.1 | 10 | 1642 | |||
XM_017025985.1 | XP_016881474.1 | 10 | 1608 | |||
XM_017025986.1 | XP_016881475.1 | 9 | 1417 | |||
XM_017025987.1 | XP_016881476.1 | 9 | 1384 | |||
GenBank mRNA | AK022604.1 | - | 10 | 1542 | ||
AK301964.1 | - | 9 | 1337 | |||
AK303117.1 | - | 9 | 1402 | |||
BC026307.1 | - | 10 | 1607 | 4792211 | ||
BC033026.1 | - | 10 | 1542 |