Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304944.1 | NP_001291873.1 | 3 | 1461 | ||
NM_024902.3 | NP_079178.2 | 2 | 999 | |||
XM_005269155.2 | XP_005269212.1 | 2 | 510 | |||
XM_005269156.2 | XP_005269213.1 | 2 | 510 | |||
XM_005269157.2 | XP_005269214.1 | 2 | 510 | |||
GenBank mRNA | AK055747.1 | - | 3 | 1461 | ||
BC033236.1 | - | 2 | 658 | 5427606 |