Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 9 | 974 | ||
NM_024919.4 | NP_079195.3 | 9 | 1124 | |||
NR_110312.1 | 13 | 1747 | ||||
XM_011536137.1 | XP_011534439.1 | 11 | 1273 | |||
XM_011536138.1 | XP_011534440.1 | 11 | 1089 | |||
XM_011536140.1 | XP_011534442.1 | 10 | 1128 | |||
XM_011536141.1 | XP_011534443.1 | 10 | 1068 | |||
XM_011536142.2 | XP_011534444.1 | 10 | 1106 | |||
XM_011536143.1 | XP_011534445.1 | 10 | 1004 | |||
XM_011536145.1 | XP_011534447.1 | 10 | 1043 | |||
XM_011536146.2 | XP_011534448.1 | 6 | 1896 | |||
XM_017011317.1 | XP_016866806.1 | 10 | 1204 | |||
XM_017011318.1 | XP_016866807.1 | 6 | 1862 | |||
GenBank mRNA | AK074110.1 | - | 10 | 2793 | ||
AK097179.1 | - | 9 | 1124 | |||
AK097579.1 | - | 9 | 974 | |||
AK125963.1 | - | 13 | 1719 | |||
AK303194.1 | - | 8 | 778 | |||
AK303407.1 | - | 8 | 901 | |||
AK316478.1 | - | 8 | 778 | |||
AL133077.1 | - | 7 | 746 | |||
BC137144.1 | - | 9 | 1082 | 9021141 |